Order Entry
Ireland
ContactUsLinkComponent
Human TROP1/EpCAM ELISA kit for Serum, Plasma, Cell Culture Supernatant and Urine. 1 * 96 items
Catalog # BIRBORB78960-96
Supplier:  Biorbyt
Human TROP1/EpCAM ELISA kit for Serum, Plasma, Cell Culture Supernatant and Urine. 1 * 96 items
Catalog # BIRBORB78960-96
Supplier:  Biorbyt