Order Entry
Ireland
ContactUsLinkComponent
PDCD4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 469 amino acids and having a m olecular mass of 51 kDa. 1 * 10 µG
Catalog # BIRBORB81263-10
Supplier:  Biorbyt
PDCD4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 469 amino acids and having a m olecular mass of 51 kDa. 1 * 10 µG
Catalog # BIRBORB81263-10
Supplier:  Biorbyt