Order Entry
Ireland
ContactUsLinkComponent
Bcl6 Human Recombinant (aa 1-150) expressed in E.coli, shows 43 kDa band on SDS-PAGE (Icluding GST tag).The Bcl6 is purified by prop rietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80959-2
Supplier:  Biorbyt
Bcl6 Human Recombinant (aa 1-150) expressed in E.coli, shows 43 kDa band on SDS-PAGE (Icluding GST tag).The Bcl6 is purified by prop rietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80959-2
Supplier:  Biorbyt