Order Entry
Ireland
ContactUsLinkComponent
L-Selectin Human Recombinant is expressed in E.coli containing 294 amino acids 39-332 fused to an amino terminal hexahistidine tag, having total molecular weight of 37.55kDa. 1 * 2 µG
Catalog # BIRBORB81201-2
Supplier:  Biorbyt
L-Selectin Human Recombinant is expressed in E.coli containing 294 amino acids 39-332 fused to an amino terminal hexahistidine tag, having total molecular weight of 37.55kDa. 1 * 2 µG
Catalog # BIRBORB81201-2
Supplier:  Biorbyt