Order Entry
Ireland
ContactUsLinkComponent
Recombinant Full-Length H3N2 A/New York/55/04 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cel ls and its Mw is 70,000 dalton. 1 * 2 µG
Catalog # BIRBORB81782-2
Supplier:  Biorbyt
Recombinant Full-Length H3N2 A/New York/55/04 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cel ls and its Mw is 70,000 dalton. 1 * 2 µG
Catalog # BIRBORB81782-2
Supplier:  Biorbyt