Order Entry
Ireland
ContactUsLinkComponent
Truncated BID Mouse Recombinant also called BH3-interacting domain death agonist p15 produced in E.coli is a single, non-glycosylate d polypeptide chain containing 61-195 amino acids (135 a.a.) and having a molecular mass of 15.4 kDa. 1 * 2 µG
Catalog # BIRBORB81409-2
Supplier:  Biorbyt
Truncated BID Mouse Recombinant also called BH3-interacting domain death agonist p15 produced in E.coli is a single, non-glycosylate d polypeptide chain containing 61-195 amino acids (135 a.a.) and having a molecular mass of 15.4 kDa. 1 * 2 µG
Catalog # BIRBORB81409-2
Supplier:  Biorbyt