Order Entry
Ireland
ContactUsLinkComponent
CD72 Human Recombinant (aa 1-98) expressed in E.coli, shows 38 kDa band on SDS-PAGE. The CD72 is purified by proprietary chromatogra phic techniques. 1 * 10 µG
Catalog # BIRBORB81013-10
Supplier:  Biorbyt
CD72 Human Recombinant (aa 1-98) expressed in E.coli, shows 38 kDa band on SDS-PAGE. The CD72 is purified by proprietary chromatogra phic techniques. 1 * 10 µG
Catalog # BIRBORB81013-10
Supplier:  Biorbyt