Order Entry
Ireland
ContactUsLinkComponent
CD146 Human Recombinant (aa 433-647) expressed in E.coli, shows 47 kDa band on SDS-PAGE. The CD146 is purified by proprietary chroma tographic techniques. 1 * 2 µG
  BIRBORB81001-2
 :  Biorbyt
CD146 Human Recombinant (aa 433-647) expressed in E.coli, shows 47 kDa band on SDS-PAGE. The CD146 is purified by proprietary chroma tographic techniques. 1 * 2 µG
  BIRBORB81001-2
 :  Biorbyt
Frequently Bought Together