Order Entry
Ireland
ContactUsLinkComponent
RAB5A Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 215 amino acids and having a m olecular mass of 23kDa. 1 * 5 µG
Catalog # BIRBORB81294-5
Supplier:  Biorbyt
RAB5A Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 215 amino acids and having a m olecular mass of 23kDa. 1 * 5 µG
Catalog # BIRBORB81294-5
Supplier:  Biorbyt