TNFR1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 162 amino acids and having to tal molecular mass of 18.2 kDa. TNFR1 Human Recombinant is purified by proprietary chromatographic technique 1 * 5 µG
Catalog # BIRBORB80623-5
Supplier: Biorbyt
Frequently Bought Together