Order Entry
Ireland
ContactUsLinkComponent
TNF -b His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 171 amino acids fragment (35-205) and having a molecular mass of 23.3kDa with an amino-terminal hexahistidine tag. The TNF-b His is 1 * 5 µG
Catalog # BIRBORB80621-5
Supplier:  Biorbyt
TNF -b His Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 171 amino acids fragment (35-205) and having a molecular mass of 23.3kDa with an amino-terminal hexahistidine tag. The TNF-b His is 1 * 5 µG
Catalog # BIRBORB80621-5
Supplier:  Biorbyt