Order Entry
Ireland
ContactUsLinkComponent
EBI3 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and having a mo lecular mass of 23 kDa. The Murine EBI3 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80297-2
Supplier:  Biorbyt
EBI3 Mouse Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and having a mo lecular mass of 23 kDa. The Murine EBI3 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80297-2
Supplier:  Biorbyt