Order Entry
Ireland
ContactUsLinkComponent
Recombinant Murine Interleukin-36 alpha, 153a.a. Interleukin-36 (IL-36) is a pro inflammatory cytokine which plays an important role in the pathophysiology of several diseases. IL-36 alpha, IL-36 beta, and IL-36 gamma (formerly IL-1F6, IL-1 1 * 2 µG
  BIRBORB107765-2
 :  Biorbyt
Recombinant Murine Interleukin-36 alpha, 153a.a. Interleukin-36 (IL-36) is a pro inflammatory cytokine which plays an important role in the pathophysiology of several diseases. IL-36 alpha, IL-36 beta, and IL-36 gamma (formerly IL-1F6, IL-1 1 * 2 µG
  BIRBORB107765-2
 :  Biorbyt
Frequently Bought Together