Order Entry
Ireland
ContactUsLinkComponent
Outlet drain tubing solvent resistant 1m. drain tubing 1/8 in id x 1/4 in od for spray chamber to peristaltic pump waste line. used with glass cyclonic spray chamber. per m 1 * 1 items
Catalog # VARI3710035400
Supplier:  VARIAN
Outlet drain tubing solvent resistant 1m. drain tubing 1/8 in id x 1/4 in od for spray chamber to peristaltic pump waste line. used with glass cyclonic spray chamber. per m 1 * 1 items
Catalog # VARI3710035400
Supplier:  VARIAN
Frequently Bought Together