Order Entry
Ireland
ContactUsLinkComponent
Variable angle cuvette holder, dra-ca-3 for cary 1/3. centre mounted. suitable for mounting a standard 1 mm pathlength cuvette insid e the sphere. 1 * 1 items
Catalog # VARI7910037500
Supplier:  VARIAN
Variable angle cuvette holder, dra-ca-3 for cary 1/3. centre mounted. suitable for mounting a standard 1 mm pathlength cuvette insid e the sphere. 1 * 1 items
Catalog # VARI7910037500
Supplier:  VARIAN
Frequently Bought Together