Order Entry
Ireland
ContactUsLinkComponent
Tubing and connectors kit - radial icp includes 2 packs of standard peristaltic pump tubing,snap connectors and other tubing used in sample introduction. ideal for spare start-up kit. 1 * 1 items
Catalog # VARI9910105400
Supplier:  VARIAN
Tubing and connectors kit - radial icp includes 2 packs of standard peristaltic pump tubing,snap connectors and other tubing used in sample introduction. ideal for spare start-up kit. 1 * 1 items
Catalog # VARI9910105400
Supplier:  VARIAN
Frequently Bought Together