Order Entry
Ireland
ContactUsLinkComponent
Sps-3 autosampler cover. 1/pk. prevents sample and standard contamination from dust and drafts. provision for optional purge or exha ust connection to remove corrisive or solvent fumes. 1 * 1 items
Catalog # VARI910245400
Supplier:  VARIAN
undefined
Sps-3 autosampler cover. 1/pk. prevents sample and standard contamination from dust and drafts. provision for optional purge or exha ust connection to remove corrisive or solvent fumes. 1 * 1 items
Catalog # VARI910245400
Supplier:  VARIAN
Frequently Bought Together