Order Entry
Ireland
ContactUsLinkComponent
Ice,,maker CM-140KE,,(Air,,cooled), Ice,,Production,,per,,24h 95-140kg, Approx Bin,,Capacity 32kg 1 * 1 items
Catalog # HOSHCM-140KE
Supplier:  HOSHIZAKI
Ice,,maker CM-140KE,,(Air,,cooled), Ice,,Production,,per,,24h 95-140kg, Approx Bin,,Capacity 32kg 1 * 1 items
Catalog # HOSHCM-140KE
Supplier:  HOSHIZAKI
Frequently Bought Together