Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

VWR®, Digital Timer, One Channel

Digital timer, single channel, White/black, ±0,01% (1 s), 63,5×12,7×57 mm
  609-0203
VWR®, Digital Timer, One Channel
  609-0203
VWR®, Digital Timer, One Channel
VWR®, Digital Timer, One Channel

Digital timer, single channel, White/black, ±0,01% (1 s), 63,5×12,7×57 mm

 
53,90 €
   
1 * 1 items
 

Timing capacity: 99 minutes, 59 seconds in steps of 1 second (±0,01%).

  • LCD
  • Counts up and down
  • Numbered keys make it simple to set countdown time
  • Alarm

 : Compliance with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001, indicating traceability to NIST.

 : Supplied with clip, stand, magnet, opening for lanyard, battery and certificate of calibration.

 
  • Colour:
    White/black
  • Description:
    Digital timer, single channel
  • W×D×H:
    63,5×12,7×57 mm
  • Accuracy:
    ±0,01% (1 s)