Order Entry
Ireland
ContactUsLinkComponent
pH combination electrode with Memosens screw plug head with Liquid electroly, NTC 30K, plastic adapter for hose connection incl. Pg1 3.5 thread, platinum junction, electrolyte L 300, sphere membrane, S-glass, length 225 mm, 12 mm Ø, +10...+135°C, 0.. 1 * 1 items
  SCOI285118210
pH combination electrode with Memosens screw plug head with Liquid electroly, NTC 30K, plastic adapter for hose connection incl. Pg1 3.5 thread, platinum junction, electrolyte L 300, sphere membrane, S-glass, length 225 mm, 12 mm Ø, +10...+135°C, 0.. 1 * 1 items
  SCOI285118210
Frequently Bought Together