Order Entry
Ireland
ContactUsLinkComponent
09.0450 Cylinder Unit LI 08.042 for Determination of Wettability in Powders, for standard measure of the time, that the powder needs, to absorb deionized water. Complete unit with stand and 4 funnels made of stainless steel. 1 * 1 items
Catalog # IBSA09.0450
Supplier:  IBS Stock Items
09.0450 Cylinder Unit LI 08.042 for Determination of Wettability in Powders, for standard measure of the time, that the powder needs, to absorb deionized water. Complete unit with stand and 4 funnels made of stainless steel. 1 * 1 items
Catalog # IBSA09.0450
Supplier:  IBS Stock Items
Frequently Bought Together