Centring rings, type KF
Centring rings with viton-O-ring, stainless steel, type KF, DN 40
Catalog # ILMV701075
Supplier: Welch by Gardner Denver
Jump to:
Product Details & Documents
Specifications
- Description:Centring rings with viton-O-ring, stainless steel, type KF
- Seq:15
- Size:DN 40