HNB Dilution Solvent, 100 ml, Used for the determination of calcium and magnesium in brine. 1 * 1 items
Catalog # HACH2305042
Supplier: Hach
HNB Dilution Solvent, 100 ml, Used for the determination of calcium and magnesium in brine. 1 * 1 items
Catalog # HACH2305042
Supplier: Hach
Frequently Bought Together
Ethanolamine ≥99%, GPR RECTAPUR® for liquid scintillation counting
Supplier: VWR Chemicals
n-Hexane ≥99%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Methanol, SupraSolv® for gas chromatography, Supelco®
Supplier: Merck
Toluene ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
Butyl acetate ≥99.5% (by GC), EMSURE® for analysis, Supelco®
Supplier: Merck
Ethanolamine, EMSURE® for analysis, Supelco®
Supplier: Merck
1,4-Dioxane 99+% stabilised ACS, meets analytical specifications of Reagent Ph. Eur.
Supplier: Thermo Fisher Scientific
Methanol 99.9% for spectroscopy
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), CHROMASOLV™ for LC-MS, Riedel-de Haën™
Supplier: Honeywell Chemicals
1-Butanol ≥98.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Ethylene Glycol (Reag. USP, Ph. Eur.) for analysis 1 * 1.000 mL
Supplier: PanReac AppliChem