Order Entry
Ireland
ContactUsLinkComponent
recombinant/native protein, ttbk2 (human) recombinant protein, human ttbk2 (np_775771.3, 1 a.a. - 331 a.a.) partial recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5658
Supplier:  Abnova
recombinant/native protein, ttbk2 (human) recombinant protein, human ttbk2 (np_775771.3, 1 a.a. - 331 a.a.) partial recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5658
Supplier:  Abnova