Order Entry
Ireland
ContactUsLinkComponent
recombinant/native protein, ret (s891a) (human) recombinant protein, human ret (np_066124.1, 658 a.a. - 1114 a.a.) s891a mutant part ial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5632
Supplier:  Abnova
recombinant/native protein, ret (s891a) (human) recombinant protein, human ret (np_066124.1, 658 a.a. - 1114 a.a.) s891a mutant part ial recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5632
Supplier:  Abnova