recombinant/native protein, irak4 (human) recombinant protein, human irak4 (aax22228.1, 1 a.a. - 460 a.a.) full-length recombinant p rotein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5566
Supplier: Abnova
Frequently Bought Together