Order Entry
Ireland
ContactUsLinkComponent
recombinant/native protein, camk2b (human) recombinant protein, human camk2b (np_742078.1, 1 a.a. - 503 a.a.) full-length recombinan t protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5504
Supplier:  Abnova
recombinant/native protein, camk2b (human) recombinant protein, human camk2b (np_742078.1, 1 a.a. - 503 a.a.) full-length recombinan t protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5504
Supplier:  Abnova