Order Entry
Ireland
ContactUsLinkComponent
recombinant/native protein, aurka (human) recombinant protein, human aurka (np_940835.1, 1 a.a. - 403 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5494
Supplier:  Abnova
recombinant/native protein, aurka (human) recombinant protein, human aurka (np_940835.1, 1 a.a. - 403 a.a.) full-length recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5494
Supplier:  Abnova