NP-40 lysis buffer, low salt
NP-40 lysis buffer, low salt, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless
Documents
Specifications
Specifications
- Pk:250 mL
- Description:NP-40 lysis buffer, low salt
- Cat. No.:J60902.AK
- Storage temperature:2°C to 8°C
- Physical form:Liquid
- Appearance:Colourless



