Order Entry
Ireland
Orders LinkContactUsLinkComponent
Stopcocks, three way, T-bore

Stopcocks, three way, T-bore

2,5 mm, 9 mm
Catalog # BIBBSST3025
Supplier:   SciLabware
Stopcocks, three way, T-bore
Catalog # BIBBSST3025
Supplier:  SciLabware
Stopcocks, three way, T-bore
Stopcocks, three way, T-bore

2,5 mm, 9 mm

Price
348,00 €
Log in for availability
1 * 5 items
Product Details & Documents

Pyrex® borosilicate glass with PTFE key

  • Replaceable NS keys with T-bore
  • Screw thread retaining device
Specifications
  • Hole Ø:
    2,5 mm
  • Seq:
    01
  • For tube O-Ø:
    9 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.