Order Entry
Ireland
ContactUsLinkComponent
10 mpos dead-end path, u-electric, RS232/485 1/4 -28*1.5mm, 75C/250psi liq, PPS/E2 Universal electric actuator Serial interface: RS-232/485 not incl.: interface cable I-22697 1 * 1 items
Catalog # VICIC25-6180EUHA
Supplier:  VICI
10 mpos dead-end path, u-electric, RS232/485 1/4 -28*1.5mm, 75C/250psi liq, PPS/E2 Universal electric actuator Serial interface: RS-232/485 not incl.: interface cable I-22697 1 * 1 items
Catalog # VICIC25-6180EUHA
Supplier:  VICI
Frequently Bought Together