Order Entry
Ireland
ContactUsLinkComponent
Standard rack for upright freezer for (hxd) 3x4 = 12 boxes 130mm high, delivered including standard white cardboard boxes and 9x9 ce ll dividers, stainless steel, dimensions (hxdxw) 401x558x140 1 * 1 items
Catalog # TENATE23334B
Supplier:  TENAK
Standard rack for upright freezer for (hxd) 3x4 = 12 boxes 130mm high, delivered including standard white cardboard boxes and 9x9 ce ll dividers, stainless steel, dimensions (hxdxw) 401x558x140 1 * 1 items
Catalog # TENATE23334B
Supplier:  TENAK
Frequently Bought Together