Alu FlexRack290 for chest freezers, aluminum, with locking rod, dimensions (hxdxw) 290x140x145mm, with 4 shelves 1 * 1 items
TENATE20104
: TENAK
Alu FlexRack290 for chest freezers, aluminum, with locking rod, dimensions (hxdxw) 290x140x145mm, with 4 shelves 1 * 1 items
TENATE20104
: TENAK
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection