Order Entry
Ireland
ContactUsLinkComponent
PRIMARY ANTIBODY, MONOCLONAL, HLA-B MONOCLONAL ANTIBODY, CLONE EP-4, MOUSE MONOCLONAL ANTIBODY RAISED AGAINST NATIVE HLA-B. 1 * 100 µG
Catalog # ABNOMAB5202
Supplier:  Abnova
PRIMARY ANTIBODY, MONOCLONAL, HLA-B MONOCLONAL ANTIBODY, CLONE EP-4, MOUSE MONOCLONAL ANTIBODY RAISED AGAINST NATIVE HLA-B. 1 * 100 µG
Catalog # ABNOMAB5202
Supplier:  Abnova