Order Entry
Ireland
ContactUsLinkComponent
KIT, THYROID PEROXIDASE IGG ELISA KIT, THYROID PEROXIDASE IGG ELISA KIT IS USED FOR THE DETECTION OF IGG ANTIBODY TO THYROID PEROXID ASE IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
  ABNOKA0952
 :  Abnova
KIT, THYROID PEROXIDASE IGG ELISA KIT, THYROID PEROXIDASE IGG ELISA KIT IS USED FOR THE DETECTION OF IGG ANTIBODY TO THYROID PEROXID ASE IN HUMAN SERUM OR PLASMA. 1 * 1 KIT
  ABNOKA0952
 :  Abnova
Frequently Bought Together