Order Entry
Ireland
ContactUsLinkComponent
KIT, IGG2C (MOUSE) ELISA KIT, IGG2C (MOUSE) ELISA KIT IS A HIGHLY SENSITIVE TWO-SITE ENZYME-LINKED IMMUNOASSAY FOR THE QUANTITATIVE DETERMINATION OF IGG2C IN MOUSE BIOLOGICAL SAMPLES. 1 * 1 KIT
Catalog # ABNOKA1970
Supplier:  Abnova
KIT, IGG2C (MOUSE) ELISA KIT, IGG2C (MOUSE) ELISA KIT IS A HIGHLY SENSITIVE TWO-SITE ENZYME-LINKED IMMUNOASSAY FOR THE QUANTITATIVE DETERMINATION OF IGG2C IN MOUSE BIOLOGICAL SAMPLES. 1 * 1 KIT
Catalog # ABNOKA1970
Supplier:  Abnova