Order Entry
Ireland
ContactUsLinkComponent
KIT, ADIPOQ (HUMAN) ELISA KIT, ADIPOQ (HUMAN) ELISA KIT IS A COMPETITIVE ENZYME-LINKED IMMUNOSORBENT ASSAY FOR THE QUANTITATIVE MEAS UREMENT OF HUMAN ADIPONECTIN IN SERUM AND PLASMA. 1 * 1 KIT
Catalog # ABNOKA0016
Supplier:  Abnova
KIT, ADIPOQ (HUMAN) ELISA KIT, ADIPOQ (HUMAN) ELISA KIT IS A COMPETITIVE ENZYME-LINKED IMMUNOSORBENT ASSAY FOR THE QUANTITATIVE MEAS UREMENT OF HUMAN ADIPONECTIN IN SERUM AND PLASMA. 1 * 1 KIT
Catalog # ABNOKA0016
Supplier:  Abnova