Milk butyrometer
Butyrometer for milk, 0 - 16% : 0,2
Catalog # 614-0096
Supplier: GERBER FUNKE
Jump to:
Product Details & Documents
Borosilicate glass.
- With or without stopper
Available with a declaration of conformity per batch and type (in German only).
Specifications
- Description:Butyrometer for milk, 0 - 16% : 0,2
- Seq:10
Accessories
Product Information
- Catalog #AvailabilityPrice
- Stopper, rubber, conical, for all measuring method butyrometersStopper, rubber, conical, for all measuring method butyrometers614-2028Log in for availability1 * 100 items - 48,20 €