Spanner wrenches
Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns, Galvanised steel
Catalog # 28-9647-76
Supplier: Cytiva
Jump to:
Product Details & Documents
These spanner wrenches are used with HiScale™ columns. To readjust the position of the adapter in the column a spanner can be used to turn the end cap. To prevent the column from rotating, a second spanner can be used to hold the end housing.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Description:Spanner wrench, for nuts Ø 58 - 62 mm, for HiScale™ 16 columns
- Material:Galvanised steel
- Seq:01