Order Entry
Ireland
ContactUsLinkComponent
Microscope stand Axio Lab.A1 HAL 35, FL-LED, 5x H - Z-drive with fine drive knob left and fine drive disk right, flat with scala - nosepiece 5x brightfield, M27 - integrated 12V DC 50W power unit, stabilized 100...240V AC/ 50...60Hz/110VA - transmi 1 * 1 items
Catalog # ZEIS4300379020000
Supplier:  ZEISS
Microscope stand Axio Lab.A1 HAL 35, FL-LED, 5x H - Z-drive with fine drive knob left and fine drive disk right, flat with scala - nosepiece 5x brightfield, M27 - integrated 12V DC 50W power unit, stabilized 100...240V AC/ 50...60Hz/110VA - transmi 1 * 1 items
Catalog # ZEIS4300379020000
Supplier:  ZEISS
Frequently Bought Together