Order Entry
Ireland
ContactUsLinkComponent
U-TTR-2 trinocular ergo tube Tilting trinocular observation tube with adjustable inclination of 5-35. Maximum field number 22 (compatible with WHN eyepieces and U-CT30-2 centering telescope). Interpupillary adjustment from 50-76mm (Siedentopf type) 1 * 1 items
Catalog # OLYMN2710000
Supplier:  OLYMPUS OPTICAL
U-TTR-2 trinocular ergo tube Tilting trinocular observation tube with adjustable inclination of 5-35. Maximum field number 22 (compatible with WHN eyepieces and U-CT30-2 centering telescope). Interpupillary adjustment from 50-76mm (Siedentopf type) 1 * 1 items
Catalog # OLYMN2710000
Supplier:  OLYMPUS OPTICAL
Frequently Bought Together