Order Entry
Ireland
ContactUsLinkComponent
Bottles glass alpha sirop amber 60ml type III soda lime glass with fitted R3/28mm black urea caps with polythene cone inserts. Bottle only height 94.0 +/- 0.8mm dia 38.6 +/- 0.7mm 1 * 40 items
Catalog # 215-2899
Supplier:  Azpack
undefined
Bottles glass alpha sirop amber 60ml type III soda lime glass with fitted R3/28mm black urea caps with polythene cone inserts. Bottle only height 94.0 +/- 0.8mm dia 38.6 +/- 0.7mm 1 * 40 items
Catalog # 215-2899
Supplier:  Azpack
Frequently Bought Together