Order Entry
Ireland
ContactUsLinkComponent
Recombinant protein, lhx4( np_203129.1, 1 a.a. - 390 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00089884-P01
Supplier:  Abnova
Recombinant protein, lhx4( np_203129.1, 1 a.a. - 390 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00089884-P01
Supplier:  Abnova