Recombinant protein, batf( np_006390.1, 1 a.a. - 125 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00010538-P01
: Abnova
Recombinant protein, batf( np_006390.1, 1 a.a. - 125 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
ABNOH00010538-P01
: Abnova
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection