Order Entry
Ireland
ContactUsLinkComponent
Recombinant protein, ifit1( np_001539.2, 1 a.a. - 478 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00003434-P01
Supplier:  Abnova
Recombinant protein, ifit1( np_001539.2, 1 a.a. - 478 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00003434-P01
Supplier:  Abnova