Order Entry
Ireland
ContactUsLinkComponent
Recombinant protein, eif4g1( aah07788, 1 a.a. - 646 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00001981-P01
Supplier:  Abnova
Recombinant protein, eif4g1( aah07788, 1 a.a. - 646 a.a.) full-length recombinant protein with gst tag. 1 * 2 µG
Catalog # ABNOH00001981-P01
Supplier:  Abnova