0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
MICROSPH.SIZE STDS 10 µM + CERTIFICAAT 1 * 15 mL
MSPP4210A
:
MARKET SOURCE PART PROCESS
MICROSPH.SIZE STDS 10 µM + CERTIFICAAT 1 * 15 mL
MSPP4210A
:
MARKET SOURCE PART PROCESS
Frequently Bought Together
Fluoride standard solution, 1,000 mg/L F- in Water (from NaF) Certipur®, Supelco®
: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
: Merck
VWR®, Conductivity standard solutions
: VWR Chemicals
pH buffer solutions
: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Nitrate standard solution, 1000 mg/L NO₃<sup>-</sup> in Water (from NaNO₃) Certipur®, Supelco®
: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Sulphuric acid 1 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1086804
: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
: Merck
Micro-weighing container inclusive 10 filters and 3 cover caps 1 * 1 items
: Brand