SYSTEM LUMINAIRE CER 236 2X36W/230V/50HZ/IP20 ALUMINIUM ANODIZED (ZO) POWER LINE: 200 MM 1 * 1 items
Catalog # WLDMCER0010UK
Supplier: WALDMANN
SYSTEM LUMINAIRE CER 236 2X36W/230V/50HZ/IP20 ALUMINIUM ANODIZED (ZO) POWER LINE: 200 MM 1 * 1 items
Catalog # WLDMCER0010UK
Supplier: WALDMANN
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection