OVS sampling tubes, ORBO™
ORBO™ 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
Catalog # 20029-U
Supplier: Merck
Jump to:
Product Details & Documents
The OSHA Versatile Sampler (OVS) is a unique tube design with the adsorbent and filter combined in one tube for collection of both vapors and aerosols.
- Packed with specially cleaned low background porous polymers
- Most common sampling rate is 1 L/min to obtain a sample volume from 60 to 480 L
It′s most common use is for sampling pesticides but can also be used for other applications.
Specifications
- Description:ORBO™ 65P (OVS) Amberlite® XAD®-4 specially cleaned, 160/80 mg RC,GFF,F,F (specially cleaned) separators, OSHA Versatile Sampler (OVS), O.D.×L: 13-8×75 mm, package of 10 ea
- Seq:01