Order Entry
Ireland
ContactUsLinkComponent
Safety cabinet with wing door. Outer carcass: 1195 x 595 x 2080 (WxDxH) mm, Inner carcass: 1099 x 446 x 1830 (WxDxH) mm, Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, Wing doors in RAL 7035 light grey, Inner carcass in RAL 7035 li 1 * 1 items
Catalog # 132-0662
Supplier:  DUPERTHAL
Safety cabinet with wing door. Outer carcass: 1195 x 595 x 2080 (WxDxH) mm, Inner carcass: 1099 x 446 x 1830 (WxDxH) mm, Outer carcass, sheet steel, powder-coated in RAL 7035 light grey, Wing doors in RAL 7035 light grey, Inner carcass in RAL 7035 li 1 * 1 items
Catalog # 132-0662
Supplier:  DUPERTHAL
Frequently Bought Together